Learn More
Eppendorf™ Centrifuge 5430 - Microcentrifuge
BIOPHARMAGREENER_CHOICE
Product Code. 10318192 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
48 x 1.5/2.0 mL, (6 x 15/50 mL, 2 x MTP)
Controller Type:
Keypad
Includes:
Without Rotor
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Controller Type Includes Voltage unitSize
10318192 48 x 1.5/2.0 mL, (6 x 15/50 mL, 2 x MTP) Keypad Without Rotor 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10318192 Supplier Eppendorf™ Supplier No. 5427000011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The unique cross-over Eppendorf Centrifuge 5430 combines the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, it can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock™ – System for quickly opening and closing the rotor lid
Order Info
Dimensions: 16.8L x 13.2W x 10 in. H (42 x 33 x 25cm), Net weight: 63.8 lb. (29kg). Most models supplied with a rotor, other rotors are available separately. Refrigerated models are available separately.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 48 x 1.5/2.0 mL, (6 x 15/50 mL, 2 x MTP)
Max. Speed 17,500 rpm
Max. RCF 30,130 x g
Display Large, Brightly Lit LCD
Height (English) Exterior 9.8 in
Height (Metric) Exterior 25 cm
Length (English) Exterior 16.8 in.
Length (Metric) Exterior 42 cm
Timer 30 sec. to 99:59 hr., Continuous Run Function
Width (English) Exterior 11.2 in.
Width (Metric) Exterior 33 cm
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Includes Without Rotor
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
Ventilated Yes
Electrical Requirements 230 V, 50/60 Hz (EU)
Dimensions (L x W x H) Exterior 34 x 42 x 25 cm (11.2 x 16.3 x 9.8 in.)
Controller Type Keypad
Wattage 475 W
Maximum Force Range 30,000 to 39,999 x g
Maximum Speed Range 17,000 to 17,999 rpm
Rotor Included No
Refrigerated No
Profile (Acceleration/Braking) Variable speed control from 100 to 17,500 rpm in 100 rpm increments
Run Time 30 sec. to 99 hr. 59 min. with HOLD
Standards EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 47 CFR FCC part 15, EN 50581, UL 61010-1, CAN/CSA-C22.2 No. 61010-1 (refer to manual for more details)
Dimensions (HWD) 10 x 13.2 x 16.8 in. (25 x 33 x 42 cm)
Net Weight 63.8 lb. (29 kg)
Program Storage 50-program memory with 5 keys for one-touch recall of routine programs
Product Line Eppendorf 5430
Voltage 230 V
Rotor Not Included