Learn More
Eppendorf™ HeatSealer S200 Accessory, Plate Adapters
Product Code. 15753155 Shop All Eppendorf Products
Change view
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15753155 PCR Plate Adapter Each
15733155 High Profile Adapter Each
15743155 Low Profile Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15753155 Supplier Eppendorf™ Supplier No. 5392070038
  • ONLINE EXCLUSIVE PRICE
    4510.00 NOK
    3985.38 NOK / Pack of 1
    Save 524.62 NOK
    11% Off
In Stock
For use with HeatSealer S200
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty
Two years
TRUSTED_SUSTAINABILITY
Specifications
Wattage 350 W
For Use With (Equipment) HeatSealer S200
Sealing Temperature Range 125°C to 200°C in 1°C increments
Product Type PCR Plate Adapter