Learn More
Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE
Product Code. 11833432 Shop All Eppendorf Products
Click to view available options
Product Type:
PCR Film
PCR Foil
Storage Film
Unit Size:
Pack of 100
Product Code. 11833432
Brand: Eppendorf™ 0030127838
Only null left
Add to basket
Add to basket
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polyester, Polypropylene
Autoclavable Autoclavable
No. per Pack 100
For Use With (Application) Hermetic sealing of PCR plates, especially recommended for low reaction volumes, best protection against evaporation
Product Type PCR Film
Certified RNase/DNase-free Yes
Temperature Range (Metric) -80°C to +100°C