Learn More
Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE
Product Code. 11518663 Shop All Eppendorf Products
Click to view available options
Product Type:
PCR Film
PCR Foil
Storage Film
Unit Size:
Pack of 100
Product Code. 11518663
Brand: Eppendorf™ 0030127870
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
No. per Pack 100 Pk.
For Use With (Application) High-strength adhesive that forms tight seals to minimize sample evaporation
Product Type Storage Film
Certified RNase/DNase-free Yes