Learn More
Eppendorf™ FA-45-6-30 Rotor Adapters
Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 5 mL Conical Tube
1 x 5 mL Tube
13 x 100 mm Tubes
13 x 75 mm Tubes
16 x 100 mm Tubes
16 x 75 mm Tubes
Unit Size:
Pack of 2
Description
- FA-45-6-30 Rotor Adapters
- For volumes up to 50mL
Specifications
Specifications
| Type | Rotor Adapter |
| Capacity | 13 x 75 mm Tubes |
| For Use With (Equipment) | Rotor for 5804/5810 centrifuge |
| Vessels per Adapter | 1 Tube |