Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE
Product Code. 10297987 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Colorless
Skirt Style:
Semi-skirted
Unskirted
Unskirted, Divisible
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
PCR Clean and Free of Bacterial DNA (Non-sterile)
Packaging:
5 Bags x 4 Plates
Individually Blister Wrapped
Unit Size:
Pack of 10
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Purity or Quality Grade Packaging unitSize
10297987 Colorless Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10176287 Colorless Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped Pack of 10
10319597 Blue Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped Pack of 10
15665367 Colorless Semi-skirted Forensic DNA Grade (Non-sterile) Individually Blister Wrapped Pack of 10
10608439 Colorless Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10441528 Blue Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10258037 Blue Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10297987 Supplier Eppendorf™ Supplier No. 0030133374
Only null left
Add to basket
Add to basket
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Colorless
Product Type PCR Plate
Skirt Style Unskirted, Divisible
Material Polypropylene, Polycarbonate
Volume (Metric) Well 250 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Ideal for extreme applications such as multiple heat sealing.
Max. RCF 2250 x g
Profile Regular
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Well Color Colorless