Learn More
Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R
Product Code. 12846213 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
20 x 5.0 mL
24 x 15 mL
4 x 250 mL
40 x 15 mL
48 x 1.5/2.0 mL
6 x 15/50 mL
Type:
Aerosol-tight Rotor
Fixed Angle Rotor
Rotor
Swinging Bucket Rotor
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Type Unit Size
12846213 4 x 250 mL Swinging Bucket Rotor Each
12886233 48 x 1.5/2.0 mL Aerosol-tight Rotor Each
15118364 20 x 5.0 mL Aerosol-tight Rotor Each
10733912 6 x 15/50 mL Aerosol-tight Rotor Each
12856243 24 x 15 mL Fixed Angle Rotor Each
12846243 40 x 15 mL Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12846213 Supplier Eppendorf™ Supplier No. 5804746007
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Rotor
Capacity 4 x 250 mL
Max. RCF 3,234 x g
Max. Speed 4,200 rpm
For Use With (Equipment) Centrifuges 5804 R and 5810 R
Includes 250 mL round buckets (x4)
Model S-4-72