Learn More
Abnova™ EPAS1 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Product Code. 16127684
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16127684 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16127684 Supplier Abnova™ Supplier No. H00002034T02.100uL
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Human overexpression lysate
Specifications
For Use With (Application) Western Blot
Description EPAS1 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_001430.3
Molecular Weight (g/mol) Theoretical M.W. 96.5kDa
Plasmid pCMV-EPAS1 full-length
Quality Control Testing Transient overexpression cell lysate was tested with anti-EPAS1 antibody (Mfr. No. H00002034-D01P) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
compliance-icons
Product Identifier
  • 100UL EPAS1 293T Cell Transient Overexpression Lys
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Mercaptoethanol