Learn More
H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers
Product Code. 13069132 Shop All SP Scienceware Products
Change view
Click to view available options
Temperature Range (Metric):
-1°C to +61° C
-10°C to +150°C
-10°C to +225°C
Length (Metric) Overall:
200 mm
350 mm
420 mm
Immersion Type:
Partial
Partial Immersion
Total
Unit Size:
Each
Product Code. Temperature Range (Metric) Length (Metric) Overall Immersion Type unitSize
13069132 -10°C to +150°C 200 mm Partial Each
13079132 -10°C to +150°C 200 mm Total Each
13009142 -10°C to +225°C 350 mm Total Each
13029142 -1°C to +61° C 420 mm Partial Immersion Each
4 options
Product Code. 13069132
Brand: H-B Instrument™ B605010100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Certified environmentally safe thermometers replace dangerous mercury.
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Specifications
Certifications/Compliance Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion Type Partial
Length (English) Overall 7.9 in.
Length (Metric) Overall 200 mm
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +150°C
Accuracy ±1°C, ±1.5°C above 110°C
Subdivision 1°C
Immersion Length 50 mm