Learn More
Memmert™ Entry Port Right (Individual Location)
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Not in combination with illumination units
- Moisture tight
- Can be closed by flap and silicone stopper
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | IPP260eco |
| Diameter (Metric) | 23 mm |