Learn More
Memmert™ Entry Port Right Centre/Top
Product Code. 17100235 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17100235 IPP410eco Each
17100195 IPP260eco Each
17120275 IPP750eco Each
17139834 HPP410eco Each
17149694 HPP110eco Each
17160065 IPP1060eco Each
17170105 IPP110eco Each
17189644 HPP1060eco Each
17189784 HPP260eco Each
17199874 HPP750eco Each
10 options
Product Code. 17100235
Brand: Memmert™ IPP410ecoF3
Only null left
Add to basket
Add to basket
For introducing connections at the side and can be closed by flap
  • Standard position right centre/top
  • Not for modelsize 110 with light module
Specifications
Color Clear
For Use With (Equipment) IPP410eco
Diameter (Metric) 23 mm