Description
- Standard position right centre/centre
- Not for model sizes 110 and 260 with light module
- Not for model size 110 with light module
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | IPP260eco |
| Diameter (Metric) | 23 mm |