Learn More
Memmert™ Entry Port Rear (Individual Position)
Product Code. p-7220919 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17119654
Brand: Memmert™ HPP1060ecoF6
Only null left
Add to basket
Add to basket
Entry port with flap closure and custom position TBC in the back
  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper
Specifications
Color Clear
For Use With (Equipment) HPP1060eco
Diameter (Metric) 23 mm