Learn More
Memmert™ Entry Port Left (Individual Location)
Product Code. 17199644 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17199644
Brand: Memmert™ HPP1060ecoF4
Only null left
Add to basket
Add to basket
Entry port flap closure custom TBC position in left side
  • Entry port flap closure custom TBC position in left side
  • Can be closed by the flap
Specifications
Color Clear
Description Not in combination with illumination units, moisture tight, can be closed by flap and silicone stopper
For Use With (Equipment) HPP1060eco
Diameter (Metric) 23 mm