Learn More
Memmert™ Entry Port Left (Individual Location)
Product Code. 17110195 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17110195 IPP260eco Each
17109884 HPP750eco Each
17110235 IPP410eco Each
17130275 IPP750eco Each
17149724 HPP1400eco Each
17149834 HPP410eco Each
17150135 IPP1400eco Each
17159694 HPP110eco Each
17160155 IPP2200eco Each
17170065 IPP1060eco Each
17180105 IPP110eco Each
17189744 HPP2200eco Each
17199644 HPP1060eco Each
17199784 HPP260eco Each
14 options
Product Code. 17110195
Brand: Memmert™ IPP260ecoF4
Only null left
Add to basket
Add to basket
Entry port flap closure custom TBC position in left side
  • Entry port flap closure custom TBC position in left side
  • Can be closed by the flap
Specifications
Color Clear
Description Can be closed by flap
For Use With (Equipment) IPP260eco
Diameter (Metric) 23 mm