Description
- Standard position left centre/top
- Not for modelsize 110 with light module
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | HPP260eco |
| Diameter (Metric) | 23 mm |