Learn More
Memmert™ Entry Port Custom Position
Product Code. 17129794 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17129794 HPP260eco Each
17100075 IPP1060eco Each
17110115 IPP110eco Each
17129654 HPP1060eco Each
17139884 HPP750eco Each
17140195 IPP260eco Each
17140235 IPP410eco Each
17160275 IPP750eco Each
17179834 HPP410eco Each
17189694 HPP110eco Each
10 options
Product Code. 17129794
Brand: Memmert™ HPP260ecoF7
Only null left
Add to basket
Add to basket
Silicone Entry Port flap and stopper closure custom position TBC in the back
  • Individual position at the back (please, state location; not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Color Clear
For Use With (Equipment) HPP260eco
Material Silicone
Diameter (Metric) 40 mm