Learn More
Memmert™ Entry Port 57 mm for UN750
Product Code. 17580396 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
57 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
17580396 57 mm Each
1 options
Product Code. 17580396
Brand: Memmert™ UN750F8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UN750 Oven
Type Entry Port
Diameter (Metric) 57 mm