Learn More
Memmert™ Entry Port 57 mm for IF110
Product Code. 17516635 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF110 Incubator
Unit Size:
Each
Product Code. 17516635
Brand: Memmert™ IF110F8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF110 Incubator
Type Entry Port
Diameter (Metric) 57 mm