Learn More
Memmert™ Entry Port 23 mm for IF110, Custom TBC Right Side
Product Code. 17586625 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17586625
Brand: Memmert™ IF110F5
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in right side
Specifications
For Use With (Equipment) IF110 Incubator
Type Entry Port
Diameter (Metric) 23 mm