Learn More
Memmert™ Entry Port 14 mm for IF110
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF110 Incubator
Unit Size:
Each
Description
- Entry port with 14 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
Specifications
| For Use With (Equipment) | IF110 Incubator |
| Type | Entry Port |
| Diameter (Metric) | 14 mm |