Learn More
Abnova™ ENTPD8 (Human) Recombinant Protein
Human ENTPD8 full-length ORF ( AAI46510.1, 1 a.a. - 458 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00377841-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGLSRKEQVFLALLGASGVSGLTALILLLVEATSVLLPTDIKFGIVFDAGSSHTSLFLYQWLANKENGTGVVSQALACQVEGPGISSYTSNAAQAGESLQGCLEEALVLIPEAQHRKTPTFLGATAGMRLLSRKNSSQARDIFAAVTQVLGRSPVDFWGAELLAGQAEGAFGWITVNYGLGTLVKYSFTGEWIQPPEEMLVGALDMGGASTQITFVPGGPILDKSTQADFRLYGSDYSVYTHSYLCFGRDQMLSRLLVGLVQSRPAALLRHPCYLSGYQTTLALGPLYESPCVHATPPLSLPQNLTVEGTGNPGACVSAIRELFNFSSCQGQEDCAFDGVYQPPLRGQFYVEASYPGQDRWLRDYCASGLYILTLLHEGYGFSEETWPSLEFRKQAGGVDIGWTLGYMLNLTGMIPADAPAQWRAESYGVWVAKVVFMVLALVAVVGAALVQLFWLQD
Specifications
AAI46510.1 | |
ectonucleoside triphosphate diphosphohydrolase 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENTPD8 | |
GST |
377841 | |
Wheat germ expression system | |
10 μg | |
GLSR2492, NTPDase-8, UNQ2492 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |