missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ENTPD8 (Human) Recombinant Protein
Human ENTPD8 full-length ORF ( AAI46510.1, 1 a.a. - 458 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAI46510.1 |
---|---|
Gene ID (Entrez) | 377841 |
Name | ectonucleoside triphosphate diphosphohydrolase 8 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130424
|
Abnova™
H00377841-P01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140424
|
Abnova™
H00377841-P01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MGLSRKEQVFLALLGASGVSGLTALILLLVEATSVLLPTDIKFGIVFDAGSSHTSLFLYQWLANKENGTGVVSQALACQVEGPGISSYTSNAAQAGESLQGCLEEALVLIPEAQHRKTPTFLGATAGMRLLSRKNSSQARDIFAAVTQVLGRSPVDFWGAELLAGQAEGAFGWITVNYGLGTLVKYSFTGEWIQPPEEMLVGALDMGGASTQITFVPGGPILDKSTQADFRLYGSDYSVYTHSYLCFGRDQMLSRLLVGLVQSRPAALLRHPCYLSGYQTTLALGPLYESPCVHATPPLSLPQNLTVEGTGNPGACVSAIRELFNFSSCQGQEDCAFDGVYQPPLRGQFYVEASYPGQDRWLRDYCASGLYILTLLHEGYGFSEETWPSLEFRKQAGGVDIGWTLGYMLNLTGMIPADAPAQWRAESYGVWVAKVVFMVLALVAVVGAALVQLFWLQD
Specifications
AAI46510.1 | |
ectonucleoside triphosphate diphosphohydrolase 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
GLSR2492, NTPDase-8, UNQ2492 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
377841 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENTPD8 | |
GST |