Learn More
Merck Millipore Vent Filters for Pure Water Storage Reservoirs
For use with Pure Water Reservoirs
1985.00 NOK - 2875.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10224372
|
Merck Millipore
TANKMPK02 |
RiOs™ 5/8/16 system |
1985.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10020272
|
Merck Millipore
TANKMPK01 |
Elix™ 3/5/10 system |
2875.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
For use with Pure Water Reservoirs
- Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
Specifications
Vent Filter | |
Polyethylene |