Learn More
Merck Millipore Vent Filters for Pure Water Storage Reservoirs
For use with Pure Water Reservoirs
Brand:  Merck Millipore TANKMPK01
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 10020272
  • 2875.00 NOK / Each
Estimated Shipment: 03-06-2024
Description
For use with Pure Water Reservoirs
  • Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
Specifications
Specifications
Polyethylene
Elix™ 3/5/10 system
Vent Filter