Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Elliptical
Product Code. 10650693 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
10 mm
15 mm
25 mm
70 mm
Diameter (Metric):
6 mm
10 mm
14 mm
28 mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 10650693
Only null left
Add to basket
Add to basket
Use for significantly increased magnetic strength and resistance to demagnetization. Rare earth stir bars are easily identified by a chemically inert carbon black spot.
  • Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
  • Samarium-cobalt core is resistant to demagnetization
  • Identified by a chemically inert, carbon black spot
The high magnetic coupling power of rare earth stir bars may cause abrasion of the PTFE coating against the base of the container. When stirring arrays are used, as in multi-well plates, lateral interaction between stir bars may be observed.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Samarium-Cobalt magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 28 mm
Length (Metric) 70 mm
Quantity 1/Ea.
Shape Elliptical