Learn More
Eppendorf™ Electronic Multipette™ BASIC-Trade-In Dispenser
GREENER_CHOICE
Product Code. p-200132155 Shop All Eppendorf Products
Click to view available options
Model:
E3
E3x
Unit Size:
Each
Product Code. 15441920
Brand: Eppendorf™ 4987000339
Creates a versatile dispensing system with one button tip ejection
  • Creates a versatile dispensing system that functions according to the positive displacement principle
  • Capable of pipetting any liquid while maintaining the highest standards for accurate results
  • High speed dispensing with motorized piston
  • Automatic Combitip™ advanced™ tip recognition
  • One button tip ejection
  • Volume range from 1μl to 50mL
  • Li-ion battery
  • Illuminated display
  • Includes 2 Combitips advanced™ assortment packs
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Lot Specific Certificate
Compatible Tips Combitips Advanced
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Natural
Disposable Reusable
For Use With CombiTip
Material Plastic
Model E3
Number of Channels 1
Programmability Step-based
Sterility Non-sterile
Type Motorized Repetitive Dispensing Pipettor
Volume (Metric) 1 μL to 50 mL
Includes E3 Multi Dispensing Range
Plug Type AU/EU/GB/US
Electrical Requirements 100/240 V, 50/60 Hz
Voltage 100/240 V