Description
- 1 mm wire, 5 mm mesh size, 5 mm frames
- equipped with 2 hinged handles
- Maximum load 10 kg
Specifications
Specifications
| Product Type | Insert, Half |
| Model | E 142 |
| Dimensions (L x W x H) | 480 x 225 x 15/55mm |
| Height (Metric) | 15/55 mm |
| Length (Metric) | 480 mm |
| Quantity | 1 Each |