missing translation for 'onlineSavingsMsg'
Learn More

Proteintech EIF4G1 Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 2B10G8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-67199-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17659876

  • 3499.43 NOK / 100µL
Estimated Shipment: 06-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EIF4G1 Monoclonal antibody apecifically detetects EIF4G1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications

Specifications

EIF4G1
Monoclonal
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
EIF4G1
EIF4G1 fusion protein Ag8357
100 μL
1981
Store at -20°C. Avoid exposure to light.
IgG1
Immunofluorescence
2B10G8
CoraLite 488
BC007788
Mouse
Protein G purification
Primary
Human, Mouse, Rat
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers