missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4G1 Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 2B10G8, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | EIF4G1 |
|---|---|
| Clone | 2B10G8 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711309
|
Proteintech
CL488-67199-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659876
|
Proteintech
CL488-67199-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EIF4G1 Monoclonal antibody apecifically detetects EIF4G1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| EIF4G1 | |
| Immunofluorescence | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 1981 | |
| EIF4G1 fusion protein Ag8357 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 2B10G8 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC007788 | |
| EIF4G1 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title