Learn More
Novus Biologicals™ EGF Phospho-Specific Array Slides
Product Code. 18604130 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Slides
Unit Size:
Pack of 2
Product Code. Quantity unitSize
18604130 2 Slides Pack of 2
1 options
Product Code. 18604130
Brand: Novus Biologicals™ AA0023
Specifications
Quantity 2 Slides
For Use With (Application) Protein Array
Sample Type Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Target Specificity EGF Phospho-Specific Array
Content And Storage Store at 4°C. Do not freeze.