missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | EEA1 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16806105
|
Proteintech
28347-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16896095
|
Proteintech
28347-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EEA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EEA1. This antibody recognizes human, mouse, rat antigen. The EEA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| EEA1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 300 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 8411 | |
| Primary |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts