Learn More
EEA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28347-1-AP-20UL
Description
The EEA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EEA1. This antibody recognizes human, mouse, rat antigen. The EEA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| EEA1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 8411 | |
| IgG |
| Western Blot, Immunofluorescence | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |