Learn More
EEA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28347-1-AP-150UL
 View more versions of this product
Product Code. 16896095
  • 4836.10 NOK / 150µL
Description
The EEA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EEA1. This antibody recognizes human, mouse, rat antigen. The EEA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EEA1
Polyclonal
Unconjugated
150 μL
8411
IgG
Western Blot, Immunofluorescence
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse