Learn More
Hanna Instruments™ Edge™ dissolved oxygen electrode with temperature sensor, HI 764080, Hanna
Product Code. 15168249
Change view
Click to view available options
Dissolved Oxygen (Saturation Range):
0 to 300%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dissolved Oxygen (Saturation Range) Unit Size
15168249 0 to 300% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15168249 Supplier Hanna Instruments™ Supplier No. HI764080
  • ONLINE EXCLUSIVE PRICE
    8631.00 NOK
    8039.22 NOK / Pack of 1
    Save 591.78 NOK
    6% Off
View all promotions
Estimated Shipment: 18-11-2026
Specifications
Length (Metric) Cable 1 m
Sensor Types Temperature Sensor
Temperature (English) Operating 32°F to 122°F
Connections 3.5 mm
Dissolved Oxygen (Saturation Range) 0 to 300%
Length (Metric) 120 mm
Type DO Electrode
Temperature (Metric) Operating 0°C to 50°C