Learn More
Tenak™ ECO AluRack for Upright Freezers, 130 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11350B
Additional Details : Weight : 1.60000kg
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
2 x 5 | |
Aluminum | |
683 x 139 x 268 mm |
Seawater resistant | |
Boxes (10) | |
Upright Freezers | |
130 mm (Box) |