Learn More
Tenak™ ECO AluRack for Upright Freezers, 130 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
1425.00 NOK - 2375.00 NOK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 130 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12412570
|
Tenak™
TE11342B |
Boxes (6) |
1425.00 NOK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
12412580
|
Tenak™
TE11352B |
Boxes (15) |
2375.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12432570
|
Tenak™
TE11344B |
Boxes (8) |
1650.00 NOK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
12452570
|
Tenak™
TE11346B |
Boxes (9) |
1725.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12472570
|
Tenak™
TE11348B |
Boxes (12) |
2035.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12492570
|
Tenak™
TE11350B |
Boxes (10) |
1880.00 NOK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Aluminum | |
130 mm (Box) |
Seawater resistant | |
Upright Freezers |