Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 85 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15610
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12498280
  • 1100.00 NOK / Each
Estimated Shipment: 23-07-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
2 x 7
Aluminum
630 x 135 x 173 mm
Microtiterplates (14)
Upright Freezers
85 mm