Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 85 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
825.00 NOK - 1540.00 NOK
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 85 mm |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
12408250
|
Tenak™
TE15514 |
Microtiterplates (20) | 4 x 5 |
1210.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12408270
|
Tenak™
TE15562 |
Microtiterplates (18) | 3 x 6 |
1210.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12408290
|
Tenak™
TE15612 |
Microtiterplates (21) | 3 x 7 |
1315.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12418270
|
Tenak™
TE15564 |
Microtiterplates (24) | 4 x 6 |
1435.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12418290
|
Tenak™
TE15614 |
Microtiterplates (28) | 4 x 7 |
1540.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12448220
|
Tenak™
TE15440 |
Microtiterplates (8) | 2 x 4 |
825.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
12458220
|
Tenak™
TE15442 |
Microtiterplates (12) | 3 x 4 |
995.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12488240
|
Tenak™
TE15510 |
Microtiterplates (10) | 2 x 5 |
880.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
12498240
|
Tenak™
TE15512 |
Microtiterplates (15) | 3 x 5 |
1050.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
12498260
|
Tenak™
TE15560 |
Microtiterplates (12) | 2 x 6 |
995.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
12498280
|
Tenak™
TE15610 |
Microtiterplates (14) | 2 x 7 |
1100.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Upright Freezers | |
85 mm |