Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 85 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE15440
Additional Details : Weight : 1.60000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
2 x 4 | |
Aluminum | |
360 x 135 x 173 mm |
Microtiterplates (8) | |
Upright Freezers | |
85 mm |