Learn More
Tenak™ ECO AluRack for Chest Freezers, 75 mm Box Height, with Boxes
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11534B
Additional Details : Weight : 1.60000kg
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Boxes (4) | |
Chest Frseezers | |
75 mm (Box) |
Seawater resistant | |
Aluminum | |
140 x 140 x 326 mm |