Learn More
Tenak™ ECO AluRack for Chest Freezers, 75 mm Box Height, with Boxes
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
790.00 NOK - 1380.00 NOK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 75 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12418140
|
Tenak™
TE11538B |
Boxes (6) |
1095.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12438130
|
Tenak™
TE11530B |
Boxes (2) |
790.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
12438140
|
Tenak™
TE11540B |
Boxes (7) |
1190.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12458130
|
Tenak™
TE11532B |
Boxes (3) |
845.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12458140
|
Tenak™
TE11542B |
Boxes (8) |
1275.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12478130
|
Tenak™
TE11534B |
Boxes (4) |
930.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12478140
|
Tenak™
TE11544B |
Boxes (9) |
1380.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12498130
|
Tenak™
TE11536B |
Boxes (5) |
1015.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Aluminum | |
75 mm (Box) |
Seawater resistant | |
Chest Frseezers |