Learn More
Tenak™ ECO AluRack for Chest Freezers, 32 mm Box Height
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
785.00 NOK - 1010.00 NOK
Specifications
Description | Seawater resistant |
---|---|
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 32 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12422580
|
Tenak™
TE11458 |
Boxes (8) |
785.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12422590
|
Tenak™
TE11466 |
Boxes (16) |
935.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12442590
|
Tenak™
TE11468 |
Boxes (18) |
975.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12462580
|
Tenak™
TE11462 |
Boxes (12) |
850.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12462590
|
Tenak™
TE11470 |
Boxes (20) |
1010.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without standard white cardboard boxes
Specifications
Seawater resistant | |
Aluminum | |
Chest Frseezers | |
32 mm (Box) |