Learn More
Tenak™ ECO AluRack for Chest Freezers, 32 mm Box Height
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11458
Additional Details : Weight : 1.60000kg
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without standard white cardboard boxes
Specifications
Seawater resistant | |
Chest Frseezers | |
140 x 140 x 280 mm |
Aluminum | |
Boxes (8) | |
32 mm (Box) |