Learn More
Invitrogen™ eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate
Product Code. 15540497
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
15540497 100 μg 100µg
1 options
Contains Sodium azide
Product Code. 15540497
Brand: Invitrogen™ 61431782
Contains Sodium azide
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.2 mg/mL
Conjugate PE-eFluor 610
Quantity 100 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PE-eFluor 610