Learn More
R&DSystems™ EAV NSP9 (NP_705590.1) VersaClone cDNA
Specifications
Specifications
| Gene | nsp9 |
| Accession Number | NP_705590.1 |
| Gene ID (Entrez) | 1489749 |
| Gene Alias | Non-structural protein 9, ORF1a polyprotein, ORF1ab polyprotein, p12 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | EAV |
| Quantity | 10 μg |