Learn More
STEMCELL Technologies EasySep™ Non-Human Primate B Cell Isolation Kit
Immunomagnetic isolation of untouched non-human primate B cells
Brand: STEMCELL Technologies 100-0345
Additional Details : Weight : 0.00440kg
Description
- The EasySep™ Non-Human Primate B Cell Isolation Kit is designed to isolate untouched and highly purified B cells from fresh or previously frozen non-human primate peripheral blood mononuclear cells (PBMCs) in as little as 20 minutes by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction
- This kit has been verified for use with rhesus and cynomolgus macaques
Specifications
B Cells | |
Cell Isolation | |
1 x 109 cells |