Learn More
STEMCELL Technologies EasySep™ Mouse Pan-Naïve T Cell Isolation Kit
Product Code. 17107971
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17107971 1 x 109 cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17107971 Supplier STEMCELL Technologies Supplier No. 19848
Immunomagnetic isolation of untouched mouse pan-naïve T cells
  • The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells (CD11b, CD19, CD24, CD25, CD44, CD45R/B220, CD49b, TER119) and streptavidin-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
Specifications
Cell Type T Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells