Learn More
STEMCELL Technologies EasySep™ Mouse Neutrophil Enrichment Kit
Description
- The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils
- Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran
- These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Granulocytes and Subsets |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |