missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Mouse CD4 Positive Selection Kit II

Product Code. 16685135
Encompass
Change view
Click to view available options
Quantity:
2 x 109 cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16685135 2 x 109 cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685135 Supplier STEMCELL Technologies Supplier No. 18952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of CD4+ T cells from mouse splenocytes or other tissues

  • The EasySep™ Mouse CD4 Positive Selection Kit II is designed to isolate CD4+ cells from single-cell suspensions of splenocytes or other tissues by positive selection
  • Desired cells are targeted with antibody complexes recognizing CD4 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product replaces the EasySep™ Mouse CD4 Positive Selection Kit (Catalog #18752) for even faster cell isolations and does not result in the labeling of isolated cells with PE

Specifications

Cell Type T Cells, T Cells, CD4+
For Use With (Application) Cell Isolation
Quantity 2 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.